missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethoxy)acetophenone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 306230050

 Afficher plus de versions de ce produit

Numéro de catalogue. AC306230050

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

4'-(Trifluoromethoxy)acetophenone
97.5
1.2780g/mL
87°C
97.5% min. (GC)
C9H7F3O2
MFCD00042404
1.278
MOEXTBIPPMLEFX-UHFFFAOYSA-N
1-[4-(trifluoromethoxy)phenyl]ethanone
522821
98%
85013-98-5
100.0
100.0°C to 102.0°C
Authentic
Glass bottle
CF3OC6H4COCH3
5g
4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
CC(=O)C1=CC=C(OC(F)(F)F)C=C1
204.15
204.15

Identifiants chimiques

85013-98-5
204.15
MOEXTBIPPMLEFX-UHFFFAOYSA-N
522821
CC(=O)C1=CC=C(OC(F)(F)F)C=C1
C9H7F3O2
MFCD00042404
4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
1-[4-(trifluoromethoxy)phenyl]ethanone
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 285-066-

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only