missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethoxy)acetophenone, 98%, Thermo Scientific™

Chemical Identifiers

CAS 85013-98-5
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00042404
InChI Key MOEXTBIPPMLEFX-UHFFFAOYSA-N
Synonym 4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
PubChem CID 522821
IUPAC Name 1-[4-(trifluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC=C(OC(F)(F)F)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC306230050
SDS View Documents
Thermo Scientific Chemicals
306230050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

85013-98-5
204.15
MOEXTBIPPMLEFX-UHFFFAOYSA-N
522821
CC(=O)C1=CC=C(OC(F)(F)F)C=C1
C9H7F3O2
MFCD00042404
4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
1-[4-(trifluoromethoxy)phenyl]ethanone

Specifications

85013-98-5
100.0
100.0°C to 102.0°C
Authentic
Glass bottle
CF3OC6H4COCH3
5g
4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
CC(=O)C1=CC=C(OC(F)(F)F)C=C1
204.15
204.15
4'-(Trifluoromethoxy)acetophenone
97.5
1.2780g/mL
87°C
97.5% min. (GC)
C9H7F3O2
MFCD00042404
1.278
MOEXTBIPPMLEFX-UHFFFAOYSA-N
1-[4-(trifluoromethoxy)phenyl]ethanone
522821
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 285-066-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only