missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcatechol, 99%

CAS: 98-29-3 | C10H14O2 | 166.22 g/mol

Supplier:  Thermo Scientific Chemicals 154350100

 Afficher plus de versions de ce produit

Numéro de catalogue. AC154350100


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

4-tert-Butylcatechol
98-29-3
100.0
1.0490g/mL
129°C
Plastic drum
(CH3)3CC6H3(OH)2
10 kg
15,54
4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
XESZUVZBAMCAEJ-UHFFFAOYSA-N
4-tert-butylbenzene-1,2-diol
7381
166.22
Technical
99%
98.5
53.0°C to 56.0°C
285.0°C
0.7% max. Catechol (GC)
C10H14O2
MFCD00002201
06,IV,6014
1.049
Solubility in water: 0.2% (25°C). Other solubilities: soluble in hot water,very sol. in alcohol,ethers,esters and ketones,partly sol. in chlorinated and aromatic hydrocarb.,slightly sol. in aliphatics. 240g/100g acetone,80g/100g benzene;60g/100g
CC(C)(C)C1=CC=C(O)C(O)=C1
166.22
90 mm2/s (25°C)
99%

Identifiants chimiques

98-29-3
166.22
XESZUVZBAMCAEJ-UHFFFAOYSA-N
7381
CC(C)(C)C1=CC=C(O)C(O)=C1
C10H14O2
MFCD00002201
4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
4-tert-butylbenzene-1,2-diol
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting eff

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-653-9

missing translation for 'rtecsNumber' : UX1400000

missing translation for 'tsca' : TSCA

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only