missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoic acid, 99%

CAS: 62-23-7 | C7H5NO4 | 167.12 g/mol

Supplier:  Thermo Scientific Chemicals A147380I

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA147380I


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

Solubility
Very slightly soluble in cold water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Spécifications

Spécifications

4-Nitrobenzoic acid
239°C to 241°C
C7H5NO4
5000 g
14,6589
Very slightly soluble in cold water.
C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]
6108
167.12
62-23-7
1.61
MFCD00007352
973593
4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova
OTLNPYWUJOZPPA-UHFFFAOYSA-N
167.12
CHEBI:262350
99%

Identifiants chimiques

62-23-7
167.12
OTLNPYWUJOZPPA-UHFFFAOYSA-N
6108
C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]
C7H5NO4
MFCD00007352
4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova
CHEBI:262350
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H301-H351-H315-H319-H335
Toxic if swallowed.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

missing translation for 'einecsNumber' : 200-526-2

missing translation for 'rtecsNumber' : DH5075000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only