missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzoic Acid 98.0+%, TCI America™

$78.93 - $637.23

Identifiants chimiques

CAS 619-84-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00002537
InChI Key YDIYEOMDOWUDTJ-UHFFFAOYSA-N
Synonym 4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid
PubChem CID 12092
IUPAC Name 4-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=C(C=C1)C(=O)O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
D072425G
Afficher les documents
TCI America
D072425G
25 g
chaque for $78.93
Il en reste null
 
D0724500G
Afficher les documents
TCI America
D0724500G
500 g
chaque for $637.23
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

619-84-1
165.192
YDIYEOMDOWUDTJ-UHFFFAOYSA-N
12092
CN(C)C1=CC=C(C=C1)C(=O)O
C9H11NO2
MFCD00002537
4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid
4-(dimethylamino)benzoic acid

Spécifications

619-84-1
C9H11NO2
25 g
YDIYEOMDOWUDTJ-UHFFFAOYSA-N
4-(dimethylamino)benzoic acid
12092
≥98.0% (HPLC,T)
4-Dimethylaminobenzoic Acid
243°C
MFCD00002537
4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid
CN(C)C1=CC=C(C=C1)C(=O)O
165.192
165.19
Crystalline Powder
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : (3)-3587

missing translation for 'rtecsNumber' : DG8735000

missing translation for 'tsca' : Yes

FDS
missing translation for 'documents'

missing translation for 'documents'