missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)benzyl Chloride 95.0+%, TCI America™
Supplier: TCI America T229025G
Spécifications
| 3-(Trifluoromethyl)benzyl Chloride | |
| Colorless | |
| C8H6ClF3 | |
| 2920 | |
| XGASTRVQNVVYIZ-UHFFFAOYSA-N | |
| 1-(chloromethyl)-3-(trifluoromethyl)benzene | |
| 69716 | |
| ≥95.0% (GC) |
| 705-29-3 | |
| 70°C | |
| 25 g | |
| 3-trifluoromethyl benzyl chloride, 1-chloromethyl-3-trifluoromethyl benzene, 3-trifluoromethylbenzyl chloride, 3-chloromethyl benzotrifluoride, m-trifluoromethylbenzyl chloride, 3-chloromethyl-benzotrifluoride, benzene, 1-chloromethyl-3-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-m-xylene, 3-chloromethylbenzotrifluoride, 3-trifluoromethylbenzylchloride | |
| C1=CC(=CC(=C1)C(F)(F)F)CCl | |
| 194.581 | |
| 194.58 | |
| Liquid |
Identifiants chimiques
| 705-29-3 | |
| 194.581 | |
| 3-trifluoromethyl benzyl chloride, 1-chloromethyl-3-trifluoromethyl benzene, 3-trifluoromethylbenzyl chloride, 3-chloromethyl benzotrifluoride, m-trifluoromethylbenzyl chloride, 3-chloromethyl-benzotrifluoride, benzene, 1-chloromethyl-3-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-m-xylene, 3-chloromethylbenzotrifluoride, 3-trifluoromethylbenzylchloride | |
| 1-(chloromethyl)-3-(trifluoromethyl)benzene |
| C8H6ClF3 | |
| XGASTRVQNVVYIZ-UHFFFAOYSA-N | |
| 69716 | |
| C1=CC(=CC(=C1)C(F)(F)F)CCl |
Sécurité et manipulation
TSCA : Yes