missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypyrazine-2-carboxamide 98.0+%, TCI America™
Supplier: TCI America H14855G
Spécifications
| 3-Hydroxypyrazine-2-carboxamide | |
| 261°C | |
| C5H5N3O2 | |
| 5 g | |
| SZPBAPFUXAADQV-UHFFFAOYSA-N | |
| 2-oxo-1H-pyrazine-3-carboxamide | |
| 294642 | |
| ≥98.0% (HPLC,T) |
| 55321-99-8 | |
| Yellow | |
| MFCD00233977 | |
| 3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry | |
| C1=CN=C(C(=O)N1)C(=O)N | |
| 139.114 | |
| 139.11 | |
| Crystalline Powder |
Identifiants chimiques
| 55321-99-8 | |
| 139.114 | |
| SZPBAPFUXAADQV-UHFFFAOYSA-N | |
| 294642 | |
| C1=CN=C(C(=O)N1)C(=O)N |
| C5H5N3O2 | |
| MFCD00233977 | |
| 3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry | |
| 2-oxo-1H-pyrazine-3-carboxamide |
Sécurité et manipulation
missing translation for 'tsca' : No