missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyrazine-2-carboxamide 98.0+%, TCI America™

Catalog No. p-7131070
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14851G 1 g
H14855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14851G Supplier TCI America Supplier No. H14851G
Only null left

Chemical Identifiers

CAS 55321-99-8
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD00233977
InChI Key SZPBAPFUXAADQV-UHFFFAOYSA-N
Synonym 3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry
PubChem CID 294642
IUPAC Name 2-oxo-1H-pyrazine-3-carboxamide
SMILES C1=CN=C(C(=O)N1)C(=O)N

Specifications

Melting Point 261°C
Color Yellow
Quantity 1 g
Formula Weight 139.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxypyrazine-2-carboxamide