missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorocatechol, 99%

CAS: 363-52-0 | C6H5FO2 | 128.10 g/mol

$278.92 - $278.92

Identifiants chimiques

CAS 363-52-0
Molecular Formula C6H5FO2
Molecular Weight (g/mol) 128.10
MDL Number MFCD00042582
InChI Key DXOSJQLIRGXWCF-UHFFFAOYSA-N
Synonym 3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142
PubChem CID 67764
ChEBI CHEBI:39876
IUPAC Name 3-fluorobenzene-1,2-diol
SMILES OC1=CC=CC(F)=C1O
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAB2214506
Afficher les documents
Thermo Scientific Chemicals
B2214506
5 g
chaque for $278.92
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

363-52-0
128.10
DXOSJQLIRGXWCF-UHFFFAOYSA-N
67764
3-fluorobenzene-1,2-diol
C6H5FO2
MFCD00042582
3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142
CHEBI:39876
OC1=CC=CC(F)=C1O

Spécifications

363-52-0
C6H5FO2
5 g
2436074
3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142
OC1=CC=CC(F)=C1O
128.10
CHEBI:39876
99%
71°C to 73°C
MFCD00042582
UN1759
Light sensitive
DXOSJQLIRGXWCF-UHFFFAOYSA-N
3-fluorobenzene-1,2-diol
67764
128.1
3-Fluorocatechol
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H334-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only