missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenol, 98%

CAS: 591-20-8 | C6H5BrO | 173.01 g/mol

$89.92 - $554.39

Identifiants chimiques

CAS 591-20-8
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.01
MDL Number MFCD00002253
InChI Key MNOJRWOWILAHAV-UHFFFAOYSA-N
Synonym m-bromophenol, phenol, 3-bromo, 3-bromo phenol, 3-bromo-phenol, phenol, m-bromo, unii-vmu0x6956y, meta bromophenol, meta-bromophenol, 3 bromophenol, 3-bromanylphenol
PubChem CID 11563
IUPAC Name 3-bromophenol
SMILES OC1=CC=CC(Br)=C1
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC146340100
FDS Afficher les documents
Thermo Scientific Chemicals
146340100
10 g Glass bottle
chaque for $89.92
Il en reste null
 
AC146340250
FDS Afficher les documents
Thermo Scientific Chemicals
146340250
25 g Glass bottle
chaque for $173.69
Il en reste null
 
AC146341000
FDS Afficher les documents
Thermo Scientific Chemicals
146341000
100 g Glass bottle
chaque for $554.39
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

591-20-8
173.01
MNOJRWOWILAHAV-UHFFFAOYSA-N
11563
OC1=CC=CC(Br)=C1
C6H5BrO
MFCD00002253
m-bromophenol, phenol, 3-bromo, 3-bromo phenol, 3-bromo-phenol, phenol, m-bromo, unii-vmu0x6956y, meta bromophenol, meta-bromophenol, 3 bromophenol, 3-bromanylphenol
3-bromophenol

Spécifications

591-20-8
100.0
Brown or Yellow
>110°C
97.5% min. (GC)
C6H5BrO
BrC6H4OH
10 g
15,1439
Solubility in water: insoluble. Other solubilities: soluble in alkalies,alcohol,ether,acetone
OC1=CC=CC(Br)=C1
173.01
173.01
Liquid
98.5
30.0°C to 32.0°C
236.0°C
Authentic
Glass bottle
1.5950 to 1.5990
MFCD00002253
06,198
m-bromophenol, phenol, 3-bromo, 3-bromo phenol, 3-bromo-phenol, phenol, m-bromo, unii-vmu0x6956y, meta bromophenol, meta-bromophenol, 3 bromophenol, 3-bromanylphenol
MNOJRWOWILAHAV-UHFFFAOYSA-N
3-bromophenol
11563
98%
3-Bromophenol
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 209-706-5

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only