Learn More
Sigma Aldrich (3-Aminopropyl)triethoxysilane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Supplier: Sigma Aldrich 440140100ML
Description
(3-Aminopropyl)tiethoxysilane (APTES) is used as an amino-silane which is mainly used as a dispersant. APTES attaches an amino group to the functional silane for bio-conjugation.Used in a preparation of a series of tenary composite films of polyimide/SiO/polydiphenylsiloxane with good optical transparency.Spécifications
| 919-30-2 | |
| 217°C (760 mmHg) | |
| H2N(CH2)3 Si(OC2H5)3 | |
| 100 mL | |
| 221.37 |
| 0.946 g/mL (at 25°C) | |
| C9H23NO3Si | |
| MFCD00008207 | |
| 3-Triethoxysilylpropylamine; APTES; APTS | |
| 99% |