missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich (3-Aminopropyl)triethoxysilane

Catalog No. p-200141978
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
Catalog No. Quantity
440140500ML 500 mL
440140100ML 100 mL
2 options

Catalog No. 440140500ML

Supplier: Sigma Aldrich 440140500ML

Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Aminopropyl)tiethoxysilane (APTES) is used as an amino-silane which is mainly used as a dispersant. APTES attaches an amino group to the functional silane for bio-conjugation.Used in a preparation of a series of tenary composite films of polyimide/SiO/polydiphenylsiloxane with good optical transparency.

Specifications

CAS 919-30-2
Density 0.946 g/mL (at 25°C)
Boiling Point 217°C (760 mmHg)
Molecular Formula C9H23NO3Si
Linear Formula H2N(CH2)3 Si(OC2H5)3
MDL Number MFCD00008207
Quantity 500 mL
Synonym 3-Triethoxysilylpropylamine; APTES; APTS
Molecular Weight (g/mol) 221.37
Percent Purity 99%