missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dehydro-L-proline 99.0+%, TCI America™
Supplier: TCI America D3825100MG
Spécifications
| 3,4-Dehydro-L-proline | |
| 217°C | |
| C5H7NO2 | |
| 100 mg | |
| OMGHIGVFLOPEHJ-BYPYZUCNSA-N | |
| (2S)-2,5-dihydro-1H-pyrrole-2-carboxylic acid | |
| 94284 | |
| ≥99.0% (HPLC) |
| 4043-88-3 | |
| White-Yellow | |
| MFCD00065964 | |
| (S)-3-Pyrroline-2-carboxylic Acid | |
| OC(=O)[C@H]1NCC=C1 | |
| 113.12 | |
| 113.12 | |
| Crystalline Powder |
Identifiants chimiques
| 4043-88-3 | |
| 113.12 | |
| OMGHIGVFLOPEHJ-BYPYZUCNSA-N | |
| 94284 | |
| OC(=O)[C@H]1NCC=C1 |
| C5H7NO2 | |
| MFCD00065964 | |
| (S)-3-Pyrroline-2-carboxylic Acid | |
| (2S)-2,5-dihydro-1H-pyrrole-2-carboxylic acid |
Sécurité et manipulation
missing translation for 'rtecsNumber' : UX9371345
missing translation for 'tsca' : No