missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dehydro-L-proline 99.0+%, TCI America™
$255.38 - $255.38
Chemical Identifiers
| CAS | 4043-88-3 |
|---|---|
| Molecular Formula | C5H7NO2 |
| Molecular Weight (g/mol) | 113.12 |
| MDL Number | MFCD00065964 |
| InChI Key | OMGHIGVFLOPEHJ-BYPYZUCNSA-N |
| Synonym | (S)-3-Pyrroline-2-carboxylic Acid |
| PubChem CID | 94284 |
| IUPAC Name | (2S)-2,5-dihydro-1H-pyrrole-2-carboxylic acid |
| SMILES | OC(=O)[C@H]1NCC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3825100MG
|
TCI America
D3825100MG |
100 mg |
N/A
|
|
|||||
Chemical Identifiers
| 4043-88-3 | |
| 113.12 | |
| OMGHIGVFLOPEHJ-BYPYZUCNSA-N | |
| 94284 | |
| OC(=O)[C@H]1NCC=C1 |
| C5H7NO2 | |
| MFCD00065964 | |
| (S)-3-Pyrroline-2-carboxylic Acid | |
| (2S)-2,5-dihydro-1H-pyrrole-2-carboxylic acid |
Specifications
| 4043-88-3 | |
| White-Yellow | |
| MFCD00065964 | |
| (S)-3-Pyrroline-2-carboxylic Acid | |
| OC(=O)[C@H]1NCC=C1 | |
| 113.12 | |
| 113.12 | |
| Crystalline Powder |
| 217°C | |
| C5H7NO2 | |
| 100 mg | |
| OMGHIGVFLOPEHJ-BYPYZUCNSA-N | |
| (2S)-2,5-dihydro-1H-pyrrole-2-carboxylic acid | |
| 94284 | |
| ≥99.0% (HPLC) | |
| 3,4-Dehydro-L-proline |
Safety and Handling
RTECSNumber : UX9371345
TSCA : No