Learn More
3,4,5-Trimethoxyaniline, 97%
CAS: 24313-88-0 | C9H13NO3 | 183.21 g/mol
$140.65 - $296.36
Identifiants chimiques
| CAS | 24313-88-0 |
|---|---|
| Molecular Formula | C9H13NO3 |
| Molecular Weight (g/mol) | 183.21 |
| MDL Number | MFCD00008393 |
| InChI Key | XEFRNCLPPFDWAC-UHFFFAOYSA-N |
| Synonym | benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline |
| PubChem CID | 32285 |
| IUPAC Name | 3,4,5-trimethoxyaniline |
| SMILES | COC1=CC(=CC(=C1OC)OC)N |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC139900100
|
Thermo Scientific Chemicals
139900100 |
10 g | Glass bottle |
chaque for $140.65
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC139900250
|
Thermo Scientific Chemicals
139900250 |
25 g | Glass bottle |
chaque for $296.36
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 24313-88-0 | |
| 183.21 | |
| XEFRNCLPPFDWAC-UHFFFAOYSA-N | |
| 32285 | |
| COC1=CC(=CC(=C1OC)OC)N |
| C9H13NO3 | |
| MFCD00008393 | |
| benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline | |
| 3,4,5-trimethoxyaniline |
Spécifications
| 24313-88-0 | |
| 100.0 | |
| Pink | |
| 97% | |
| C9H13NO3 | |
| MFCD00008393 | |
| 13, 826 | |
| Solubility in water: insoluble | |
| COC1=CC(=CC(=C1OC)OC)N | |
| 183.21 | |
| 183.21 | |
| Powder |
| 96.0 | |
| 110.0°C to 113.0°C | |
| Authentic | |
| Glass bottle | |
| (CH3O)3C6H2NH2 | |
| 10 g | |
| benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline | |
| XEFRNCLPPFDWAC-UHFFFAOYSA-N | |
| 3,4,5-trimethoxyaniline | |
| 32285 | |
| 97% | |
| 3, 4, 5-Trimethoxyaniline |
Sécurité et manipulation
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 246-154-4
RUO – Research Use Only