missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Sulfobenzoic Anhydride 97.0+%, TCI America™
Supplier: TCI America S0124100G
Spécifications
| 2-Sulfobenzoic Anhydride | |
| 125°C | |
| C7H4O4S | |
| 100 g | |
| NCYNKWQXFADUOZ-UHFFFAOYSA-N | |
| 1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one | |
| 65729 | |
| ≥97.0% (T) |
| 81-08-3 | |
| White-Yellow | |
| MFCD00005879 | |
| 2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w | |
| C1=CC=C2C(=C1)C(=O)OS2(=O)=O | |
| 184.165 | |
| 184.17 | |
| Crystalline Powder |
Identifiants chimiques
| 81-08-3 | |
| 184.165 | |
| NCYNKWQXFADUOZ-UHFFFAOYSA-N | |
| 65729 | |
| C1=CC=C2C(=C1)C(=O)OS2(=O)=O |
| C7H4O4S | |
| MFCD00005879 | |
| 2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w | |
| 1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one |
Sécurité et manipulation
missing translation for 'einecsNumber' : (5)-3565
missing translation for 'tsca' : Yes