missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzoic Anhydride 97.0+%, TCI America™

Catalog No. p-7135824
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S012425G 25 g
S0124100G 100 g
S0124500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S012425G Supplier TCI America Supplier No. S012425G
Only null left

Chemical Identifiers

CAS 81-08-3
Molecular Formula C7H4O4S
Molecular Weight (g/mol) 184.165
MDL Number MFCD00005879
InChI Key NCYNKWQXFADUOZ-UHFFFAOYSA-N
Synonym 2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w
PubChem CID 65729
IUPAC Name 1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C1=CC=C2C(=C1)C(=O)OS2(=O)=O

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Formula Weight 184.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Sulfobenzoic Anhydride
EINECSNumber (5)-3565
TSCA Yes