Learn More
2% Nitric Acid Matrix Blank, SPEX CertiPrep™
Nitric Acid Matrix Blank for ICP-MS, 125 mL in 2% HNO3, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Supplier: SPEX CertiPrep Group LLC CLBLKHNO3
Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Nitric Acid Matrix Blank
- 125 mL Volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Spécifications
| 2% Nitric Acid Blank | |
| 7697-37-2 | |
| HNO3 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 63.01 | |
| 944 |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00011349 | |
| Certified Reference Material | |
| O[N+]([O-])=O | |
| nitric acid | |
| 125 mL | |
| CHEBI:48107 |
Identifiants chimiques
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
| HNO3 | |
| MFCD00011349 | |
| 944 | |
| nitric acid |
Sécurité et manipulation
Causes skin irritation, serious eye irritation
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only