missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanol, 99%, mixture of cis and trans

CAS: 583-59-5 | C7H14O | 114.19 g/mol

Supplier:  Thermo Scientific Chemicals 166890025

 Afficher plus de versions de ce produit

Numéro de catalogue. AC166890025


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

2-Methylcyclohexanol
98.5
-38.0°C
0.9300g/mL
58°C
98.5% min. (GC)
C7H14O
CH3C6H10OH
2.5 kg
08,303
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
NDVWOBYBJYUSMF-UHFFFAOYSA-N
2-methylcyclohexan-1-ol
11418
99%
583-59-5
100.0
Colorless
163.0°C to 166.0°C
Authentic
Glass bottle
1.4615 to 1.4635
MFCD00001443
06, 11
0.93
Solubility in water: slightly soluble
CC1CCCCC1O
114.19
114.19
Liquid

Identifiants chimiques

583-59-5
114.19
NDVWOBYBJYUSMF-UHFFFAOYSA-N
11418
CC1CCCCC1O
C7H14O
MFCD00001443
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
2-methylcyclohexan-1-ol
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 209-512-

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only