missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanol, 99%, mixture of cis and trans

CAS: 583-59-5 | C7H14O | 114.19 g/mol

$111.43 - $533.96

Identifiants chimiques

CAS 583-59-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00001443
InChI Key NDVWOBYBJYUSMF-UHFFFAOYSA-N
Synonym 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
PubChem CID 11418
IUPAC Name 2-methylcyclohexan-1-ol
SMILES CC1CCCCC1O
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AC166892500
FDS Afficher les documents
Thermo Scientific Chemicals
166892500
250 g
chaque for $111.43
Il en reste null
 
AC166890010
FDS Afficher les documents
Thermo Scientific Chemicals
166890010
1 kg
chaque for $244.20
Il en reste null
 
AC166890025
FDS Afficher les documents
Thermo Scientific Chemicals
166890025
2.5 kg
chaque for $533.96
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

583-59-5
114.19
NDVWOBYBJYUSMF-UHFFFAOYSA-N
11418
CC1CCCCC1O
C7H14O
MFCD00001443
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
2-methylcyclohexan-1-ol

Spécifications

583-59-5
100.0
Colorless
163.0°C to 166.0°C
Authentic
Glass bottle
1.4615 to 1.4635
MFCD00001443
06, 11
0.93
Solubility in water: slightly soluble
CC1CCCCC1O
114.19
114.19
Liquid
98.5
-38.0°C
0.9300g/mL
58°C
98.5% min. (GC)
C7H14O
CH3C6H10OH
250 g
08,303
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
NDVWOBYBJYUSMF-UHFFFAOYSA-N
2-methylcyclohexan-1-ol
11418
99%
2-Methylcyclohexanol
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 209-512-

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only