missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-butene 98.0+%, TCI America™
Supplier: TCI America M0174100ML
Spécifications
| 2-Methyl-1-butene | |
| Colorless | |
| C5H10 | |
| 100 mL | |
| 2-methyl-1-butene, 2-methylbutene, 1-butene, 2-methyl, 1-isoamylene, 2-methylbutene-1, butene, 2-methyl, gamma-isoamylene, .gamma.-isoamylene, unii-33c9y0i55h, c2h5c ch3 =ch2 | |
| CCC(C)=C | |
| 70.14 | |
| CHEBI:77915 | |
| ≥98.0% (GC) |
| 563-46-2 | |
| 31°C | |
| MFCD00009333 | |
| 2459 | |
| MHNNAWXXUZQSNM-UHFFFAOYSA-N | |
| 2-methylbut-1-ene | |
| 11240 | |
| 70.14 | |
| Liquid |
Identifiants chimiques
| 563-46-2 | |
| 70.14 | |
| MHNNAWXXUZQSNM-UHFFFAOYSA-N | |
| 11240 | |
| 2-methylbut-1-ene |
| C5H10 | |
| MFCD00009333 | |
| 2-methyl-1-butene, 2-methylbutene, 1-butene, 2-methyl, 1-isoamylene, 2-methylbutene-1, butene, 2-methyl, gamma-isoamylene, .gamma.-isoamylene, unii-33c9y0i55h, c2h5c ch3 =ch2 | |
| CHEBI:77915 | |
| CCC(C)=C |
Sécurité et manipulation
missing translation for 'einecsNumber' : (2)-0019
missing translation for 'tsca' : Yes
Recommended Storage : Refrigerator