missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-butene 98.0+%, TCI America™

Catalog No. p-7132036
Change view
Click to view available options
Quantity:
25 mL
100 mL
Catalog No. Quantity
M017425ML 25 mL
M0174100ML 100 mL
2 options

Catalog No. M017425ML

Supplier: TCI America M017425ML

Only null left

Chemical Identifiers

CAS 563-46-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009333
InChI Key MHNNAWXXUZQSNM-UHFFFAOYSA-N
Synonym 2-methyl-1-butene, 2-methylbutene, 1-butene, 2-methyl, 1-isoamylene, 2-methylbutene-1, butene, 2-methyl, gamma-isoamylene, .gamma.-isoamylene, unii-33c9y0i55h, c2h5c ch3 =ch2
PubChem CID 11240
ChEBI CHEBI:77915
IUPAC Name 2-methylbut-1-ene
SMILES CCC(C)=C

Specifications

Color Colorless
Boiling Point 31°C
Quantity 25 mL
UN Number 2459
Formula Weight 70.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-butene
EINECSNumber (2)-0019
TSCA Yes
Recommended Storage Refrigerator