missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzamide, 98%

CAS: 529512-81-0 | C8H8FNO2 | 169.16 g/mol

Supplier:  Thermo Scientific Chemicals H3277406

 Afficher plus de versions de ce produit

Numéro de catalogue. AAH3277406


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

2-Fluoro-6-methoxybenzamide
190°C to 194°C
MFCD00277474
2-fluoro-6-methoxy-benzamide, 2-methoxy-6-fluorobenzamide, 2-fluoro-6-methoxy benzamide, #, benzamide, 2-fluoro-6-methoxy
COC1=C(C(N)=O)C(F)=CC=C1
169.16
169.15
529512-81-0
C8H8FNO2
5 g
BDJNFURQDZEPLK-UHFFFAOYSA-N
2-fluoro-6-methoxybenzamide
579738
98%

Identifiants chimiques

529512-81-0
169.16
BDJNFURQDZEPLK-UHFFFAOYSA-N
579738
COC1=C(C(N)=O)C(F)=CC=C1
C8H8FNO2
MFCD00277474
2-fluoro-6-methoxy-benzamide, 2-methoxy-6-fluorobenzamide, 2-fluoro-6-methoxy benzamide, #, benzamide, 2-fluoro-6-methoxy
2-fluoro-6-methoxybenzamide
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only