missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-D-phenylalanine, 98%

CAS: 80126-50-7 | C9H10ClNO2 | 199.63 g/mol

Supplier:  Thermo Scientific Chemicals H2735903

 Afficher plus de versions de ce produit

Numéro de catalogue. AAH2735903


Il en reste null

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

211°C to 213°C
80126-50-7
MFCD00077920
CVZZNRXMDCOHBG-UHFFFAOYNA-N
(2R)-2-amino-3-(2-chlorophenyl)propanoic acid
199.63
199.64
2-Chloro-D-phenylalanine
C9H10ClNO2
2-chloro-d-phenylalanine, h-d-phe 2-cl-oh, d-2-chlorophenylalanine, 2r-2-amino-3-2-chlorophenyl propanoic acid, d-2-chlorophe, d-phenylalanine, 2-chloro, r-2-amino-3-2-chlorophenyl propanoic acid, pubchem11960, pubchem17991, d-2-chloro phenylalanine
NC(CC1=CC=CC=C1Cl)C(O)=O
1 g
6951092
98%

Identifiants chimiques

80126-50-7
199.63
CVZZNRXMDCOHBG-UHFFFAOYNA-N
6951092
NC(CC1=CC=CC=C1Cl)C(O)=O
C9H10ClNO2
MFCD00077920
2-chloro-d-phenylalanine, h-d-phe 2-cl-oh, d-2-chlorophenylalanine, 2r-2-amino-3-2-chlorophenyl propanoic acid, d-2-chlorophe, d-phenylalanine, 2-chloro, r-2-amino-3-2-chlorophenyl propanoic acid, pubchem11960, pubchem17991, d-2-chloro phenylalanine
(2R)-2-amino-3-(2-chlorophenyl)propanoic acid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

TSCA : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only