missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-D-phenylalanine, 98%

Catalog No. p-7029847
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27359MD 250 mg
AAH2735903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27359MD Supplier Thermo Scientific Chemicals Supplier No. H27359MD

CAS: 80126-50-7 | C9H10ClNO2 | 199.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80126-50-7
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00077920
InChI Key CVZZNRXMDCOHBG-UHFFFAOYNA-N
Synonym 2-chloro-d-phenylalanine, h-d-phe 2-cl-oh, d-2-chlorophenylalanine, 2r-2-amino-3-2-chlorophenyl propanoic acid, d-2-chlorophe, d-phenylalanine, 2-chloro, r-2-amino-3-2-chlorophenyl propanoic acid, pubchem11960, pubchem17991, d-2-chloro phenylalanine
PubChem CID 6951092
IUPAC Name (2R)-2-amino-3-(2-chlorophenyl)propanoic acid
SMILES NC(CC1=CC=CC=C1Cl)C(O)=O

Specifications

Melting Point 211°C to 213°C
Quantity 250 mg
Formula Weight 199.64
Percent Purity 98%
Chemical Name or Material 2-Chloro-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only