Learn More
2-Bromobenzothiazole, 99%
CAS: 2516-40-7 | C7H4BrNS | 214.08 g/mol
Supplier: Thermo Scientific Chemicals H6328606
Description
2-Bromobenzothiazole, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromobenzothiazole, is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids and bases.
Spécifications
| 2-Bromobenzothiazole | |
| 39°C to 43°C | |
| >110°C (230°F) | |
| MFCD02681887 | |
| 2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou | |
| DRLMMVPCYXFPEP-UHFFFAOYSA-N | |
| 2-bromo-1,3-benzothiazole | |
| 612040 | |
| 99% |
| 2516-40-7 | |
| Yellow | |
| C7H4BrNS | |
| 5 g | |
| Slightly soluble in water. | |
| C1=CC=C2C(=C1)N=C(S2)Br | |
| 214.08 | |
| 214.08 | |
| Crystalline Solid |
Identifiants chimiques
| 2516-40-7 | |
| 214.08 | |
| DRLMMVPCYXFPEP-UHFFFAOYSA-N | |
| 612040 | |
| C1=CC=C2C(=C1)N=C(S2)Br |
| C7H4BrNS | |
| MFCD02681887 | |
| 2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou | |
| 2-bromo-1,3-benzothiazole |
Sécurité et manipulation
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only