missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzothiazole, 99%

CAS: 2516-40-7 | C7H4BrNS | 214.08 g/mol

$91.63 - $792.62

Chemical Identifiers

CAS 2516-40-7
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD02681887
InChI Key DRLMMVPCYXFPEP-UHFFFAOYSA-N
Synonym 2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
PubChem CID 612040
IUPAC Name 2-bromo-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6328603
View Documents
Thermo Scientific Chemicals
H6328603
1 g
Each for $91.63
Only null left
 
AAH6328606
View Documents
Thermo Scientific Chemicals
H6328606
5 g
Each for $222.42
Only null left
 
AAH6328614
View Documents
Thermo Scientific Chemicals
H6328614
25 g
Each for $792.62
Only null left
 
Description

Description

2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids and bases.
Specifications

Chemical Identifiers

2516-40-7
214.08
DRLMMVPCYXFPEP-UHFFFAOYSA-N
612040
C1=CC=C2C(=C1)N=C(S2)Br
C7H4BrNS
MFCD02681887
2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
2-bromo-1,3-benzothiazole

Specifications

2516-40-7
Yellow
C7H4BrNS
1 g
Slightly soluble in water.
C1=CC=C2C(=C1)N=C(S2)Br
214.08
214.08
Crystalline Solid
39°C to 43°C
>110°C (230°F)
MFCD02681887
2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
DRLMMVPCYXFPEP-UHFFFAOYSA-N
2-bromo-1,3-benzothiazole
612040
99%
2-Bromobenzothiazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only