missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9-fluorenone 98.0+%, TCI America™
Supplier: TCI America B310925G
Spécifications
| 2-Bromo-9-fluorenone | |
| 147°C | |
| C13H7BrO | |
| 25 g | |
| MTCARZDHUIEYMB-UHFFFAOYSA-N | |
| 2-bromo-9H-fluoren-9-one | |
| 725831 | |
| ≥98.0% (GC) |
| 3096-56-8 | |
| Yellow | |
| MFCD00010789 | |
| 2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f | |
| BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1 | |
| 259.10 | |
| 259.10 | |
| Crystalline Powder |
Identifiants chimiques
| 3096-56-8 | |
| 259.10 | |
| MTCARZDHUIEYMB-UHFFFAOYSA-N | |
| 725831 | |
| BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1 |
| C13H7BrO | |
| MFCD00010789 | |
| 2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f | |
| 2-bromo-9H-fluoren-9-one |
Sécurité et manipulation
missing translation for 'tsca' : No