missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzoic acid, 99%

CAS: 2148-56-3 | C7H6ClNO2 | 171.58 g/mol

$223.45 - $223.45

Identifiants chimiques

CAS 2148-56-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00051530
InChI Key SZCPTRGBOVXVCA-UHFFFAOYSA-N
Synonym 6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b
PubChem CID 75071
IUPAC Name 2-amino-6-chlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)C(=O)O)N
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC300500100
FDS Afficher les documents
Thermo Scientific Chemicals
300500100
10 g Glass bottle
chaque for $223.45
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

2148-56-3
171.58
SZCPTRGBOVXVCA-UHFFFAOYSA-N
75071
C1=CC(=C(C(=C1)Cl)C(=O)O)N
C7H6ClNO2
MFCD00051530
6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b
2-amino-6-chlorobenzoic acid

Spécifications

2148-56-3
100.0
Yellow to Beige
98.5% min. (HPLC)
C7H6ClNO2
10 g
SZCPTRGBOVXVCA-UHFFFAOYSA-N
2-amino-6-chlorobenzoic acid
75071
99%
2-Amino-6-chlorobenzoic acid, 99%
98.5
143°C to 147°C
Authentic
Glass bottle
MFCD00051530
6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b
C1=CC(=C(C(=C1)Cl)C(=O)O)N
171.58
171.58
Crystalline Powder
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 218-416-8

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only