missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylphenol, 99.8%

CAS: 128-37-0 | C15H24O | 220.35 g/mol

Supplier:  Thermo Scientific Chemicals 219830050

 Afficher plus de versions de ce produit

Numéro de catalogue. AC219830050


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

2, 6-Di-tert-butyl-4-methylphenol
128-37-0
100.0
White
127°C
99.8% min. (GC)
C15H24O
MFCD00011644
06,III,2073
15,155
Solubility in water: insoluble. Other solubilities: freely soluble in toluene,soluble in methanol,ethanol,isopropanol,acetone,benzene,methyl ethyl ketone,petroleum ether,,cellowolve and most other hydrocarbon solvents
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
220.35
3.47 mm2/s (0°C)
220.35
Crystals
p.a.
99.75
69.0°C to 71.0°C
265.0°C
Authentic
Plastic drum
[(CH3)3C]2C6H2(CH3)OH
5 kg
15,113
2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
NLZUEZXRPGMBCV-UHFFFAOYSA-N
2,6-ditert-butyl-4-methylphenol
31404
CHEBI:34247
99.8%

Identifiants chimiques

128-37-0
220.35
NLZUEZXRPGMBCV-UHFFFAOYSA-N
31404
2,6-ditert-butyl-4-methylphenol
C15H24O
MFCD00011644
2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
CHEBI:34247
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 204-881-4

missing translation for 'rtecsNumber' : GO7875000

missing translation for 'tsca' : TSCA

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only