Learn More
2,2-Dimethyl-1,3-dioxan-5-one, tech. 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L2017703
Description
2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.
Solubility
Miscible with water and most organic solvents.
Notes
Incompatible with oxidizing agents. Air, moisture and heat sensitive.
Spécifications
| 2,2-Dimethyl-1,3-dioxan-5-one | |
| 40°C to 46°C (14 mmHg) | |
| 1.4315 | |
| 1 g | |
| Air and moisture sensitive | |
| Miscible with water and most organic solvents. | |
| CC1(OCC(=O)CO1)C | |
| 130.143 | |
| 130.15 | |
| Technical |
| 74181-34-3 | |
| C6H10O3 | |
| MFCD00671514 | |
| 3061933 | |
| 2,2-dimethyl-1,3-dioxane-5-one, 2,2-dimethyl-1,3 dioxan-5-one, 1,3-dioxan-5-one, 2,2-dimethyl, acmc-1bg7h, ksc496q3n, 5-oxo-2,2-dimethyl-1,3-dioxane, 2,2-dimethyl-1,3-dioxan-5-one, technical 250mg | |
| ASFQDNDZFGFMMP-UHFFFAOYSA-N | |
| 2,2-dimethyl-1,3-dioxan-5-one | |
| 2733141 | |
| 90% |
Identifiants chimiques
| 74181-34-3 | |
| 130.143 | |
| ASFQDNDZFGFMMP-UHFFFAOYSA-N | |
| 2733141 | |
| CC1(OCC(=O)CO1)C |
| C6H10O3 | |
| MFCD00671514 | |
| 2,2-dimethyl-1,3-dioxane-5-one, 2,2-dimethyl-1,3 dioxan-5-one, 1,3-dioxan-5-one, 2,2-dimethyl, acmc-1bg7h, ksc496q3n, 5-oxo-2,2-dimethyl-1,3-dioxane, 2,2-dimethyl-1,3-dioxan-5-one, technical 250mg | |
| 2,2-dimethyl-1,3-dioxan-5-one |
Sécurité et manipulation
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C; Store under Argon; Store under Nitrogen
RUO – Research Use Only