Learn More
2,2-Dimethyl-1,3-dioxan-5-one, tech. 90%, Thermo Scientific Chemicals
$330.11 - $330.11
Chemical Identifiers
| CAS | 74181-34-3 |
|---|---|
| Molecular Formula | C6H10O3 |
| Molecular Weight (g/mol) | 130.143 |
| MDL Number | MFCD00671514 |
| InChI Key | ASFQDNDZFGFMMP-UHFFFAOYSA-N |
| Synonym | 2,2-dimethyl-1,3-dioxane-5-one, 2,2-dimethyl-1,3 dioxan-5-one, 1,3-dioxan-5-one, 2,2-dimethyl, acmc-1bg7h, ksc496q3n, 5-oxo-2,2-dimethyl-1,3-dioxane, 2,2-dimethyl-1,3-dioxan-5-one, technical 250mg |
| PubChem CID | 2733141 |
| IUPAC Name | 2,2-dimethyl-1,3-dioxan-5-one |
| SMILES | CC1(OCC(=O)CO1)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL2017703
|
Thermo Scientific Chemicals
L2017703 |
1 g |
Each for $330.11
|
|
|||||
Description
2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.
Solubility
Miscible with water and most organic solvents.
Notes
Incompatible with oxidizing agents. Air, moisture and heat sensitive.
Chemical Identifiers
| 74181-34-3 | |
| 130.143 | |
| ASFQDNDZFGFMMP-UHFFFAOYSA-N | |
| 2733141 | |
| CC1(OCC(=O)CO1)C |
| C6H10O3 | |
| MFCD00671514 | |
| 2,2-dimethyl-1,3-dioxane-5-one, 2,2-dimethyl-1,3 dioxan-5-one, 1,3-dioxan-5-one, 2,2-dimethyl, acmc-1bg7h, ksc496q3n, 5-oxo-2,2-dimethyl-1,3-dioxane, 2,2-dimethyl-1,3-dioxan-5-one, technical 250mg | |
| 2,2-dimethyl-1,3-dioxan-5-one |
Specifications
| 74181-34-3 | |
| C6H10O3 | |
| MFCD00671514 | |
| 3061933 | |
| 2,2-dimethyl-1,3-dioxane-5-one, 2,2-dimethyl-1,3 dioxan-5-one, 1,3-dioxan-5-one, 2,2-dimethyl, acmc-1bg7h, ksc496q3n, 5-oxo-2,2-dimethyl-1,3-dioxane, 2,2-dimethyl-1,3-dioxan-5-one, technical 250mg | |
| ASFQDNDZFGFMMP-UHFFFAOYSA-N | |
| 2,2-dimethyl-1,3-dioxan-5-one | |
| 2733141 | |
| 90% | |
| 2,2-Dimethyl-1,3-dioxan-5-one |
| 40°C to 46°C (14 mmHg) | |
| 1.4315 | |
| 1 g | |
| Air and moisture sensitive | |
| Miscible with water and most organic solvents. | |
| CC1(OCC(=O)CO1)C | |
| 130.143 | |
| 130.15 | |
| Technical |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C; Store under Argon; Store under Nitrogen
RUO – Research Use Only