Learn More
1-Bromononane, 99%
CAS: 693-58-3 | C9H19Br | 207.155 g/mol
Supplier: Thermo Scientific Chemicals A1057722
Description
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.
Solubility
Insoluble in water. Soluble in ethyl ether and benzene.
Notes
Stable under normal conditions. Incompatible with oxidizing agents.
Spécifications
| 1-Bromononane | |
| 1.086 | |
| 90°C (194°F) | |
| 1.455 | |
| 100 g | |
| n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163 | |
| AYMUQTNXKPEMLM-UHFFFAOYSA-N | |
| 1-bromononane | |
| 12742 | |
| 99% |
| 693-58-3 | |
| 200°C to 201°C | |
| C9H19Br | |
| MFCD00000278 | |
| 1733632 | |
| Insoluble in water. Soluble in ethyl ether and benzene. | |
| CCCCCCCCCBr | |
| 207.155 | |
| 207.16 |
Identifiants chimiques
| 693-58-3 | |
| 207.155 | |
| AYMUQTNXKPEMLM-UHFFFAOYSA-N | |
| 12742 | |
| CCCCCCCCCBr |
| C9H19Br | |
| MFCD00000278 | |
| n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163 | |
| 1-bromononane |
Sécurité et manipulation
GHS H Statement
H227
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H332-H335
missing translation for 'einecsNumber' : 211-755-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only