missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals
$195.95 - $195.95
Identifiants chimiques
| CAS | 15397-15-6 |
|---|---|
| Molecular Formula | C34H28O9 |
| Molecular Weight (g/mol) | 580.589 |
| MDL Number | MFCD07369658 |
| InChI Key | QJZSLTLDMBDKOU-VBHQRPIPSA-N |
| Synonym | 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-b-d-ribofuranose, 2s,3r,4r,5r-5-benzoyloxy methyl-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate, 2s,3r,4r,5r-2,4-bis benzoyloxy-5-benzoyloxy methyl-3-methyloxolan-3-yl benzoate, 1,2,3,5-tetra-o-benzoyl-2'-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d ribofuranose, 2-c-methyl-beta-d-ribofuranose 1,2,3,5-tetrabenzoate, 2-c-methyl-1,2,3,5-tetra-o-benzoyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofura |
| PubChem CID | 10875560 |
| IUPAC Name | [(2R,3R,4R,5S)-3,4,5-tribenzoyloxy-4-methyloxolan-2-yl]methyl benzoate |
| SMILES | CC1(C(C(OC1OC(=O)C2=CC=CC=C2)COC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAJ6693006
|
Thermo Scientific Chemicals
J6693006 |
5 g |
chaque for $195.95
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 15397-15-6 | |
| 580.589 | |
| QJZSLTLDMBDKOU-VBHQRPIPSA-N | |
| 10875560 | |
| CC1(C(C(OC1OC(=O)C2=CC=CC=C2)COC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5 |
| C34H28O9 | |
| MFCD07369658 | |
| 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-b-d-ribofuranose, 2s,3r,4r,5r-5-benzoyloxy methyl-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate, 2s,3r,4r,5r-2,4-bis benzoyloxy-5-benzoyloxy methyl-3-methyloxolan-3-yl benzoate, 1,2,3,5-tetra-o-benzoyl-2'-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d ribofuranose, 2-c-methyl-beta-d-ribofuranose 1,2,3,5-tetrabenzoate, 2-c-methyl-1,2,3,5-tetra-o-benzoyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofura | |
| [(2R,3R,4R,5S)-3,4,5-tribenzoyloxy-4-methyloxolan-2-yl]methyl benzoate |
Spécifications
| 15397-15-6 | |
| White | |
| MFCD07369658 | |
| 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-b-d-ribofuranose, 2s,3r,4r,5r-5-benzoyloxy methyl-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate, 2s,3r,4r,5r-2,4-bis benzoyloxy-5-benzoyloxy methyl-3-methyloxolan-3-yl benzoate, 1,2,3,5-tetra-o-benzoyl-2'-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d ribofuranose, 2-c-methyl-beta-d-ribofuranose 1,2,3,5-tetrabenzoate, 2-c-methyl-1,2,3,5-tetra-o-benzoyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofura | |
| QJZSLTLDMBDKOU-VBHQRPIPSA-N | |
| CC1(C(C(OC1OC(=O)C2=CC=CC=C2)COC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5 | |
| 580.589 | |
| 580.58 | |
| Powder |
| 147°C to 163°C | |
| C34H28O9 | |
| 5 g | |
| Soluble in chloroform or acetonitrile | |
| +70° (c=1 in Chloroform) | |
| [(2R,3R,4R,5S)-3,4,5-tribenzoyloxy-4-methyloxolan-2-yl]methyl benzoate | |
| 10875560 | |
| 98% | |
| 1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose |
Sécurité et manipulation
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only