missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals

Catalog No. aaj6693006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6693006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6693006 Supplier Thermo Scientific Chemicals Supplier No. J6693006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15397-15-6 | Molecular Formula: C34H28O9 | Molecular Weight: 580.589 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15397-15-6
Molecular Formula C34H28O9
Molecular Weight (g/mol) 580.589
MDL Number MFCD07369658
InChI Key QJZSLTLDMBDKOU-VBHQRPIPSA-N
Synonym 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-b-d-ribofuranose, 2s,3r,4r,5r-5-benzoyloxy methyl-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate, 2s,3r,4r,5r-2,4-bis benzoyloxy-5-benzoyloxy methyl-3-methyloxolan-3-yl benzoate, 1,2,3,5-tetra-o-benzoyl-2'-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d ribofuranose, 2-c-methyl-beta-d-ribofuranose 1,2,3,5-tetrabenzoate, 2-c-methyl-1,2,3,5-tetra-o-benzoyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofura
PubChem CID 10875560
IUPAC Name [(2R,3R,4R,5S)-3,4,5-tribenzoyloxy-4-methyloxolan-2-yl]methyl benzoate
SMILES CC1(C(C(OC1OC(=O)C2=CC=CC=C2)COC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5

Specifications

Melting Point 147°C to 163°C
Color White
Quantity 5 g
Solubility Information Soluble in chloroform or acetonitrile
Optical Rotation +70° (c=1 in Chloroform)
Formula Weight 580.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only