accessibility menu, dialog, popup

UserName

CAS RN 928-49-4

IUPAC Name: hex-3-yne
Synonyms: 3-hexyne
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: DQQNMIPXXNPGCV-UHFFFAOYSA-N
SMILES: CCC#CCC
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)