missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyne, 99%

Catalog No. AC308120250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC308120250 25 g Glass bottle
AC308120050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC308120250 Supplier Thermo Scientific Chemicals Supplier No. 308120250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 928-49-4 | C6H10 | 82.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 928-49-4
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00009381
InChI Key DQQNMIPXXNPGCV-UHFFFAOYSA-N
Synonym 3-hexyne, diethylacetylene, unii-9gtq990q4k, acmc-209rhw, c2h5c.$.cc2h5
PubChem CID 13568
IUPAC Name hex-3-yne
SMILES CCC#CCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7200g/mL
Boiling Point 81°C to 82°C
Flash Point -14°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH2C∽CCH2CH3
Refractive Index 1.4100 to 1.412
Quantity 25 g
Specific Gravity 0.72
Formula Weight 82.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Hexyne, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-173-4
TSCA TSCA

RUO – Research Use Only