missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyne 97.0+%, TCI America™

Catalog No. H04305ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04305ML 5 mL
H043025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04305ML Supplier TCI America Supplier No. H04305ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 928-49-4
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00009381
InChI Key DQQNMIPXXNPGCV-UHFFFAOYSA-N
Synonym 3-hexyne, diethylacetylene, unii-9gtq990q4k, acmc-209rhw, c2h5c.$.cc2h5
PubChem CID 13568
IUPAC Name hex-3-yne
SMILES CCC#CCC

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 mL
UN Number 3295
Formula Weight 82.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hexyne
TSCA No
Recommended Storage Refrigerator