missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyne, 99%

Catalog No. AAL0268214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0268214 25 g
AAL0268206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0268214 Supplier Thermo Scientific Chemicals Supplier No. L0268214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 928-49-4 | C6H10 | 82.15 g/mol

3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.

Chemical Identifiers

CAS 928-49-4
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00009381
InChI Key DQQNMIPXXNPGCV-UHFFFAOYSA-N
Synonym 3-hexyne, diethylacetylene, unii-9gtq990q4k, acmc-209rhw, c2h5c.$.cc2h5
PubChem CID 13568
IUPAC Name hex-3-yne
SMILES CCC#CCC

Specifications

Density 0.721
Boiling Point 80°C to 82°C
Flash Point −10°C (14°F)
Odor Characteristic
Refractive Index 1.411
Linear Formula CH3CH2C≡CCH2CH3
Quantity 25 g
UN Number UN3295
Beilstein 1731158
Solubility Information Not miscible in water.
Formula Weight 82.15
Percent Purity 99%
Chemical Name or Material 3-Hexyne
Show More Show Less
Hazard Category H225-H304-H315-H319-H335
Hazard Statement GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 213-173-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only