accessibility menu, dialog, popup

UserName

(R)(R)(R)(S)(R)(R)(S)(R)(R)(R)(R)(S)(S)(R)(R)OOHOHOOHOO∗HOHOOHHOHHOO∗HOHHO∗HO∗A

CAS RN 9057-02-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C18H30O15)AHO
InChi Key: —
SMILES: *.O-*.[H][C@]1(-*)O[C@H](CO)[C@@H](O[C@@]2([H])O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO-*)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)