missing translation for 'onlineSavingsMsg'
Learn More

Pullulan, TCI America™

Catalog No. P097825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P097825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P097825G Supplier TCI America Supplier No. P097825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9057-02-7
Molecular Formula (C18H30O15)AHO
MDL Number MFCD00081940
InChI Key GFZFEWWPMNSVBS-WVZDODFGSA-N
PubChem CID 131636581
IUPAC Name (2R,4S,5S)-2-[(3S,4R,6R)-6-[(3S,4R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(methoxymethyl)oxane-3,4,5-triol
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC)CO)CO)O)O)O

Specifications

Color White-Yellow
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Pullulan
RTECSNumber UO5470000
TSCA Yes