missing translation for 'onlineSavingsMsg'
Learn More

Pullulan Standard, Certified According to DIN, for GPC, 12,000, MilliporeSigma™ Supelco™

Catalog No. 111003867
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003867 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111003867 Supplier Merck Emd Millipore Supplier No. 9787325MG

Pullulan Standards are mostly used to obtain molecular mass distribution of cellulosic samples. It mainly consists of polymaltotriose units linked together by α-(1→6) linkages.

Specifications

Physical Form Neat
Type Certified Reference Material
Quantity 25 mg
Grade GPC