missing translation for 'onlineSavingsMsg'
Learn More

Pullulan Standard, Set Mp ∼350 - 700000, MilliporeSigma™ Supelco™

Catalog No. 111015872
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015872 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015872 Supplier Merck Emd Millipore Supplier No. 963511KT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pullulan Standards are mostly used to obtain molecular mass distribution of cellulosic samples. It mainly consists of polymaltotriose units linked together by α-(1→6) linkages.

Specifications

CAS 9057-02-7
Physical Form Neat
Synonym Pullulan Standard Set
Type Analytical Standard
Quantity 1 Ea.
Grade GPC