CAS RN 9004-67-5
Methyl Cellulose (Powder/USP/FCC), Fisher Chemical
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N Synonym: Cellulose Methyl Ether PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl cellulose, viscosity 1600 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cP (2% solution in water)
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl cellulose, viscosity 15 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl cellulose, viscosity 400 cP (2% solution in water)
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 8000 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 400 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 15 cP (2% solution in water)
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl Cellulose (13-18mPa.s, 2% in Water at 20°C), TCI America™
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl Cellulose (350-550mPa.s, 2% in Water at 20°C), TCI America™
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl Cellulose (1,000-1,800mPa.s, 2% in Water at 20°C), TCI America™
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl Cellulose (7,000-10,000mPa.s, 2% in Water at 20°C), TCI America™
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl cellulose, MP Biomedicals™
CAS: 9004-67-5 Molecular Formula: C20H38O11 MDL Number: MFCD00081763 Synonym: Cellulose methyl ether,Methocel MC,Cellothyl,Syncelose PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl Cellulose, viscosity of 2% aqueous solution (25°C) 15cps, MP Biomedicals
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N Synonym: Cellulose methyl ether PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
- 1
- 2