missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cellulose (20-30mPa.s, 2% in Water at 20°C), TCI America™

Catalog No. M0291500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0291500G 500 g
M029125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0291500G Supplier TCI America Supplier No. M0291500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Color White
Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material Methyl Cellulose (20-30mPa.s, 2% in Water at 20°C)
EINECSNumber (8)-0186
RTECSNumber FJ5959000
TSCA Yes