missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cellulose (3,500-5,600mPa.s, 2% in Water at 20°C), TCI America™

Catalog No. M018525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M018525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M018525G Supplier TCI America Supplier No. M018525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent grade methyl cellulose, supplied as a white to yellow powder.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Color White-Yellow
Quantity 25 g
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Methyl Cellulose (3,500-5,600mPa.s, 2% in Water at 20°C)
EINECSNumber (8)-0186
RTECSNumber FJ5959000
TSCA Yes