accessibility menu, dialog, popup

UserName

NaOOH2C

CAS RN 9003-04-7

IUPAC Name: sodium prop-2-enoate
Synonyms: polymerized acrylic acid
Molecular Weight (g/mol): 94.05
Molecular Formula: C3H3NaO2
InChi Key: NNMHYFLPFNGQFZ-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C=C
Grade 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results