missing translation for 'onlineSavingsMsg'
Learn More

Polyacrylic Acid Sodium Salt, 45% Solution, Spectrum™ Chemical

Catalog No. 18605291
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18605291 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18605291 Supplier Spectrum Chemical Mfg Cor Supplier No. P1704100GM

P1704, 9003-04-7, [CH2CH(CO2Na)]n

Spectrum™ Chemical Polyacrylic Acid Sodium Salt, 45 Percent Solution, also known as PAAc or Acrysol, is an anionic polymer used in disposable diapers and as thickening and emulsifying agents in the pharmaceutical and cosmetic industries. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9003-04-7 , 7732-18-5
Molecular Formula C3H3NaO2
Molecular Weight (g/mol) 94.05
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
IUPAC Name sodium prop-2-enoate
SMILES [Na+].[O-]C(=O)C=C

Specifications

Physical Form Viscous liquid
CAS Max % 0.45, 0.55
Quantity 100 g
Grade Reagent
Packaging Amber Glass Bottle
Viscosity 350-1000 cSt