missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid sodium salt) for GPC 83,000, MilliporeSigma™ Supelco™

Catalog No. 111015776
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015776 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015776 Supplier Merck Emd Millipore Supplier No. 81131250MG

Poly(acrylic acid sodium salt) is a hydrophilic polymer, which can find wide applications in the production of consumer productslike diapers, because of its ability to absorb many more times its mass in water.

Specifications

CAS 9003-04-7
Physical Form Neat
MDL Number MFCD00147949
Synonym Poly(sodium acrylate); Sodium polyacrylate
Type Analytical Standard
Quantity 250 mg
Grade GPC