missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid sodium salt) for GPC 16,000, MilliporeSigma™ Supelco™

Catalog No. 111015775
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015775 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015775 Supplier Merck Emd Millipore Supplier No. 81123250MG

Specifications

CAS 9003-04-7
Physical Form Neat
MDL Number MFCD00147949
Synonym Poly(sodium acrylate); Sodium polyacrylate
Type Analytical Standard
Quantity 250 mg
Grade GPC